siRNA Details
Gene Symbol: SSR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204526.1 | NP_001191455.1 | 5 | 454 | ||
NM_001204527.1 | NP_001191456.1 | 5 | 477 | |||
NM_006280.2 | NP_006271.1 | 4 | 427 | |||
NR_037927.1 | 6 | 648 | ||||
XM_011531186.1 | XP_011529488.1 | 5 | 336 | |||
XM_011531187.1 | XP_011529489.1 | 5 | 336 | |||
XM_017029756.1 | XP_016885245.1 | 5 | 1225 | |||
XM_017029757.1 | XP_016885246.1 | 5 | 960 | |||
GenBank mRNA | AK057117.1 | - | 3 | 2010 | ||
AK094395.1 | - | 4 | 2130 | |||
AK290493.1 | - | 4 | 333 | |||
BC003371.1 | - | 4 | 307 | 2900758 | ||
BE791319.1 | - | 4 | 349 | |||
BG165852.1 | - | 5 | 461 | |||
BP202705.1 | - | 5 | 416 | |||
BT007192.1 | - | 4 | 303 | |||
CD672615.1 | - | 5 | 519 | |||
DB238237.1 | - | 4 | 426 | |||
X90583.1 | - | 4 | 333 | |||
Z69043.1 | - | 4 | 352 |
