siRNA Details
Gene Symbol: TIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022037.2 | NP_071320.2 | 11 | 1175 | ||
NM_022173.2 | NP_071505.2 | 12 | 1208 | |||
XM_005264527.1 | XP_005264584.1 | 11 | 1182 | |||
XM_005264528.4 | XP_005264585.1 | 11 | 1120 | |||
XM_005264531.1 | XP_005264588.1 | 13 | 1255 | |||
XM_011533077.1 | XP_011531379.1 | 12 | 1215 | |||
XM_011533078.1 | XP_011531380.1 | 11 | 4881 | |||
XM_011533081.2 | XP_011531383.1 | 12 | 1981 | |||
XM_011533082.2 | XP_011531384.1 | 10 | 5553 | |||
XM_017004789.1 | XP_016860278.1 | 12 | 1037 | |||
XM_017004790.1 | XP_016860279.1 | 11 | 1003 | |||
XM_017004791.1 | XP_016860280.1 | 10 | 1087 | |||
XM_017004792.1 | XP_016860281.1 | 11 | 3392 | |||
XM_017004793.1 | XP_016860282.1 | 11 | 3217 | |||
XM_017004794.1 | XP_016860283.1 | 11 | 1949 | |||
XM_017004795.1 | XP_016860284.1 | 13 | 1215 | |||
XM_017004796.1 | XP_016860285.1 | 11 | 4723 | |||
XM_017004797.1 | XP_016860286.1 | 12 | 2519 | |||
XM_017004798.1 | XP_016860287.1 | 12 | 1250 | |||
XM_017004799.1 | XP_016860288.1 | 12 | 1212 | |||
XM_017004800.1 | XP_016860289.1 | 11 | 1179 | |||
GenBank mRNA | AB209211.1 | - | 12 | 1349 | ||
DR157578.1 | - | 6 | 547 | |||
M77142.1 | - | 11 | 1014 |
