siRNA Details
Gene Symbol: TIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022037.2 | NP_071320.2 | 5, 6 | 585 | ||
NM_022173.2 | NP_071505.2 | 6, 7 | 618 | |||
XM_005264527.1 | XP_005264584.1 | 5, 6 | 595 | |||
XM_005264528.4 | XP_005264585.1 | 5, 6 | 530 | |||
XM_005264531.1 | XP_005264588.1 | 7, 8 | 665 | |||
XM_011533077.1 | XP_011531379.1 | 6, 7 | 628 | |||
XM_017004789.1 | XP_016860278.1 | 6, 7 | 447 | |||
XM_017004790.1 | XP_016860279.1 | 5, 6 | 413 | |||
XM_017004791.1 | XP_016860280.1 | 4, 5 | 497 | |||
XM_017004795.1 | XP_016860284.1 | 7, 8 | 625 | |||
XM_017004799.1 | XP_016860288.1 | 6, 7 | 622 | |||
XM_017004800.1 | XP_016860289.1 | 5, 6 | 589 | |||
GenBank mRNA | AB209211.1 | - | 6, 7 | 629 | ||
BC015944.1 | - | 6, 7 | 425 | 3909177 | ||
CN295707.1 | - | 5, 6 | 501 | |||
CX867570.1 | - | 6, 7 | 591 | |||
M77142.1 | - | 5, 6 | 424 |
