siRNA Details
Gene Symbol: UROS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000375.2 | NP_000366.1 | 5 | 554 | ||
NM_001324036.1 | NP_001310965.1 | 5 | 554 | |||
NM_001324037.1 | NP_001310966.1 | 5 | 554 | |||
NM_001324038.1 | NP_001310967.1 | 5 | 554 | |||
NM_001324039.1 | NP_001310968.1 | 5 | 554 | |||
NR_136675.1 | 3 | 373 | ||||
NR_136676.1 | 5 | 554 | ||||
NR_136677.1 | 5 | 554 | ||||
NR_136678.1 | 2 | 284 | ||||
XM_005270140.4 | XP_005270197.1 | 5 | 396 | |||
XM_011540127.1 | XP_011538429.1 | 5 | 396 | |||
XM_017016610.1 | XP_016872099.1 | 5 | 396 | |||
XM_017016611.1 | XP_016872100.1 | 5 | 352 | |||
XM_017016612.1 | XP_016872101.1 | 5 | 396 | |||
GenBank mRNA | AF230665.1 | - | 4 | 421 | ||
AK314896.1 | - | 5 | 411 | |||
BC002573.2 | - | 5 | 513 | 3160463 | ||
BI668388.1 | - | 2 | 224 | |||
J03824.1 | - | 5 | 484 | |||
KU178377.1 | - | 4 | 288 | |||
KU178378.1 | - | 4 | 288 |
