siRNA Details
Gene Symbol: DYRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003583.3 | NP_003574.1 | 2 | 905 | ||
NM_006482.2 | NP_006473.2 | 3 | 1054 | |||
XM_017020032.1 | XP_016875521.1 | 2 | 1197 | |||
GenBank mRNA | AK313937.1 | - | 3 | 1066 | ||
BC005809.2 | - | 2 | 859 | 3831977 | ||
BC006375.2 | - | 4 | 865 | 4139392 | ||
Y13493.1 | - | 3 | 812 |
