siRNA Details
Gene Symbol: SMARCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003601.3 | NP_003592.3 | 14, 15 | 2376 | ||
GenBank mRNA | AB010882.1 | - | 14, 15 | 2116 | ||
AK302841.1 | - | 15, 16 | 2011 | |||
AK316444.1 | - | 15, 16 | 2178 | |||
BC023144.1 | - | 14, 15 | 2127 | 4340921 |
