siRNA Details
Gene Symbol: SMARCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003601.3 | NP_003592.3 | 4, 5 | 970 | ||
GenBank mRNA | AB010882.1 | - | 4, 5 | 710 | ||
AK302841.1 | - | 5, 6 | 605 | |||
AK308424.1 | - | 4, 5 | 700 | |||
AK316444.1 | - | 5, 6 | 772 | |||
BC023144.1 | - | 4, 5 | 721 | 4340921 |
