siRNA Details
Gene Symbol: RAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015885.1 | NP_001015885.1 | 5, 6 | 743 | ||
NM_003610.3 | NP_003601.1 | 5, 6 | 719 | |||
XM_005260582.2 | XP_005260639.2 | 5, 6 | 502 | |||
XM_005260583.2 | XP_005260640.1 | 5, 6 | 508 | |||
XM_011529087.2 | XP_011527389.1 | 5, 6 | 502 | |||
XM_011529088.2 | XP_011527390.1 | 5, 6 | 874 | |||
XM_011529089.1 | XP_011527391.1 | 5, 6 | 635 | |||
XM_017028108.1 | XP_016883597.1 | 5, 6 | 839 | |||
GenBank mRNA | AK292247.1 | - | 5, 6 | 775 | ||
AK307966.1 | - | 5, 6 | 582 | |||
AK309069.1 | - | 4, 5 | 528 | |||
AK309743.1 | - | 5, 6 | 555 | |||
AY336091.1 | - | 4, 5 | 396 | |||
AY349350.1 | - | 5, 6 | 743 | |||
BC103754.1 | - | 5, 6 | 560 | 6501538 | ||
BC106923.1 | - | 5, 6 | 517 | 40032941 | ||
BC106924.1 | - | 5, 6 | 517 | 40032942 | ||
U84720.1 | - | 5, 6 | 565 |
