siRNA Details
Gene Symbol: SNX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003794.3 | NP_003785.1 | 14 | 1359 | ||
NR_073435.1 | 13 | 1237 | ||||
XM_017007414.1 | XP_016862903.1 | 15 | 1447 | |||
GenBank mRNA | AF065485.1 | - | 15 | 1317 | ||
AF130078.1 | - | 9 | 726 | |||
AK001835.1 | - | 14 | 1321 | |||
AK298972.1 | - | 13 | 1204 | |||
BC018762.1 | - | 14 | 1335 | 4641014 |
