siRNA Details
Gene Symbol: USP34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014709.3 | NP_055524.3 | 51 | 6614 | ||
GenBank mRNA | AB011142.2 | - | 52 | 6669 | ||
AJ586138.1 | - | 49 | 6139 | |||
AK023845.1 | - | 4 | 480 | |||
AK125898.1 | - | 10 | 1133 | |||
AL831918.1 | - | 13 | 1426 |
