siRNA Details
Gene Symbol: KIAA0232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100590.1 | NP_001094060.1 | 6 | 2590 | ||
NM_014743.2 | NP_055558.2 | 7 | 2674 | |||
XM_005248041.4 | XP_005248098.1 | 4 | 2149 | |||
XM_011513598.2 | XP_011511900.1 | 7 | 2327 | |||
XM_017008860.1 | XP_016864349.1 | 6 | 3620 | |||
XM_017008861.1 | XP_016864350.1 | 5 | 2337 | |||
XM_017008862.1 | XP_016864351.1 | 6 | 2975 | |||
XM_017008863.1 | XP_016864352.1 | 5 | 2337 | |||
GenBank mRNA | AK124715.1 | - | 6 | 2530 | ||
BC150286.1 | - | 7 | 2666 | 8860111 | ||
D86985.3 | - | 7 | 2666 | |||
EF553521.1 | - | 6 | 2531 |
