siRNA Details
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 16 | 2733 | ||
XM_006721612.2 | XP_006721675.1 | 14 | 2176 | |||
XM_011524090.2 | XP_011522392.1 | 17 | 2799 | |||
XM_011524091.2 | XP_011522393.1 | 17 | 4184 | |||
XM_011524095.2 | XP_011522397.1 | 17 | 2531 | |||
XM_011524096.1 | XP_011522398.1 | 17 | 2799 | |||
XM_011524098.2 | XP_011522400.1 | 15 | 2430 | |||
XM_011524099.2 | XP_011522401.1 | 15 | 1920 | |||
XM_011524100.2 | XP_011522402.1 | 14 | 1702 | |||
XM_017025455.1 | XP_016880944.1 | 17 | 2742 | |||
XM_017025456.1 | XP_016880945.1 | 14 | 2494 | |||
XM_017025457.1 | XP_016880946.1 | 13 | 2264 | |||
GenBank mRNA | AB018296.1 | - | 16 | 2478 | ||
AK292876.1 | - | 16 | 2495 | |||
AK296971.1 | - | 15 | 2375 | |||
AK300547.1 | - | 9 | 881 | |||
AL080108.1 | - | 10 | 1166 | |||
BC017566.1 | - | 16 | 2492 | |||
BC113016.1 | - | 15 | 2448 | 40036441 | ||
BC113017.1 | - | 14 | 2341 | 40036446 |
