siRNA Details
Gene Symbol: FCHSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014824.2 | NP_055639.2 | 6 | 727 | ||
XM_011545409.1 | XP_011543711.1 | 6 | 609 | |||
XM_011545410.2 | XP_011543712.1 | 6 | 542 | |||
XM_017018632.1 | XP_016874121.1 | 6 | 786 | |||
GenBank mRNA | AB018312.1 | - | 5 | 577 | ||
AK296376.1 | - | 5 | 397 | |||
AK297928.1 | - | 6 | 611 | |||
AL133567.1 | - | 1 | 95 | |||
BC010394.1 | - | 6 | 474 | 4091973 | ||
CD358961.1 | - | 6 | 741 |
