siRNA Details
Gene Symbol: RHOBTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242359.1 | NP_001229288.1 | 12 | 2457 | ||
NM_014836.4 | NP_055651.1 | 11 | 2385 | |||
NR_024554.2 | 12 | 2259 | ||||
NR_024555.1 | 12 | 2223 | ||||
XM_006718084.1 | XP_006718147.1 | 11 | 2193 | |||
XM_006718085.1 | XP_006718148.1 | 11 | 2220 | |||
XM_011540424.1 | XP_011538726.1 | 12 | 2251 | |||
XM_011540425.1 | XP_011538727.1 | 11 | 2145 | |||
XM_017017000.1 | XP_016872489.1 | 11 | 2440 | |||
XM_017017001.1 | XP_016872490.1 | 11 | 2436 | |||
XM_017017002.1 | XP_016872491.1 | 10 | 2330 | |||
XM_017017003.1 | XP_016872492.1 | 13 | 2326 | |||
XM_017017004.1 | XP_016872493.1 | 13 | 2328 | |||
XM_017017005.1 | XP_016872494.1 | 12 | 2233 | |||
XM_017017006.1 | XP_016872495.1 | 12 | 2282 | |||
XM_017017007.1 | XP_016872496.1 | 10 | 2332 | |||
XM_017017008.1 | XP_016872497.1 | 12 | 2493 | |||
GenBank mRNA | AB018283.3 | - | 11 | 2276 | ||
AK075129.1 | - | 12 | 2223 | |||
AK307630.1 | - | 10 | 1955 | |||
AK309440.1 | - | 3 | 321 | |||
BC032848.1 | - | 12 | 2434 | 4830471 | ||
BC041791.1 | - | 11 | 2176 | 5297756 |
