siRNA Details
Gene Symbol: AKAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005858.3 | NP_005849.1 | 10 | 1251 | ||
XM_011527624.2 | XP_011525926.1 | 10 | 1344 | |||
XM_017026141.1 | XP_016881630.1 | 10 | 1396 | |||
GenBank mRNA | AK303640.1 | - | 7 | 878 | ||
AK308251.1 | - | 9 | 1145 | |||
AK309529.1 | - | 7 | 779 | |||
AL050160.1 | - | 4 | 179 | |||
BC037270.2 | - | 10 | 1241 | 4837309 | ||
Y11997.1 | - | 10 | 1242 |
