siRNA Details
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199573.1 | NP_001186502.1 | 4, 5 | 1121 | ||
NM_006074.4 | NP_006065.2 | 4, 5 | 1133 | |||
GenBank mRNA | AK290434.1 | - | 4, 5 | 863 | ||
AK298715.1 | - | 5, 6 | 813 | |||
AK298934.1 | - | 4, 5 | 687 | |||
AK301192.1 | - | 3, 4 | 604 | |||
AK308796.1 | - | 3, 4 | 753 | |||
AL360134.1 | - | 2, 3 | 291 | |||
AM040752.1 | - | 1, 2 | 81 | |||
BC022281.1 | - | 4, 5 | 940 | 4718024 | ||
BC035582.1 | - | 4, 5 | 906 | 5583800 | ||
HQ842635.1 | - | 3, 4 | 754 | |||
X82200.1 | - | 4, 5 | 864 |
