siRNA Details
Gene Symbol: TGOLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206840.1 | NP_001193769.1 | 2 | 1433 | ||
NM_001206841.1 | NP_001193770.1 | 2 | 1433 | |||
NM_001206844.1 | NP_001193773.1 | 3 | 1259 | |||
NM_006464.3 | NP_006455.2 | 2 | 1433 | |||
GenBank mRNA | AF027515.1 | - | 2 | 1161 | ||
AF027516.1 | - | 2 | 1161 | |||
AK126465.1 | - | 4 | 656 | |||
AK222829.1 | - | 2 | 1125 | |||
AK223063.1 | - | 2 | 1124 | |||
AK312479.1 | - | 2 | 1126 | |||
BC008461.1 | - | 2 | 1126 | 4272702 | ||
BC028219.1 | - | 2 | 1122 | 5217823 | ||
BX640868.1 | - | 3 | 1215 | |||
U62390.1 | - | 2 | 1161 | |||
X94333.1 | - | 2 | 1117 |
