siRNA Details
Gene Symbol: IFT27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177701.2 | NP_001171172.1 | 3, 4 | 599 | ||
NM_006860.4 | NP_006851.1 | 3, 4 | 596 | |||
XM_006724106.2 | XP_006724169.1 | 4, 5 | 360 | |||
XM_017028540.1 | XP_016884029.1 | 3, 4 | 603 | |||
GenBank mRNA | AA809074.1 | - | 4, 5 | 502 | ||
AB209424.1 | - | 3, 4 | 591 | |||
AL022729.1 | - | 3, 4 | 540 | |||
BC000566.2 | - | 3, 4 | 577 | 3163341 | ||
BM711121.1 | - | 3, 4 | 187 | |||
BT006815.1 | - | 3, 4 | 173 | |||
CR456558.1 | - | 3, 4 | 215 |
