siRNA Details
Gene Symbol: LILRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130917.2 | NP_001124389.2 | 3 | 343 | ||
NM_001290270.1 | NP_001277199.1 | 2 | 307 | |||
NM_001290271.1 | NP_001277200.1 | 4 | 782 | |||
NM_006866.3 | NP_006857.2 | 4 | 915 | |||
XM_006722986.1 | XP_006723049.1 | 4 | 915 | |||
XM_011526385.1 | XP_011524687.1 | 4 | 915 | |||
XM_011526386.1 | XP_011524688.1 | 3 | 879 | |||
XM_011526387.1 | XP_011524689.1 | 3 | 879 | |||
XM_011526388.1 | XP_011524690.1 | 3 | 879 | |||
XM_011526390.1 | XP_011524692.1 | 4 | 915 | |||
XM_011526391.1 | XP_011524693.1 | 4 | 915 | |||
XM_017026224.1 | XP_016881713.1 | 4 | 915 | |||
GenBank mRNA | AF025531.1 | - | 3 | 334 | ||
AK130170.1 | - | 3 | 373 | |||
AK222651.1 | - | 3 | 339 | |||
AK310088.1 | - | 4 | 764 | |||
BC017412.1 | - | 3 | 345 | 4618777 | ||
BC027916.1 | - | 3 | 328 | 5223154 | ||
BP296918.1 | - | 3 | 384 | |||
EU915609.1 | - | 2 | 276 | |||
EU915610.1 | - | 3 | 312 | |||
U70665.1 | - | 3 | 344 | |||
U82275.1 | - | 3 | 382 | |||
U82276.1 | - | 2 | 297 | |||
U82277.1 | - | 3 | 295 | |||
U82278.1 | - | 3 | 295 |
