siRNA Details
Gene Symbol: ZSWIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242487.1 | NP_001229416.1 | 21, 22 | 4912 | ||
NM_001242488.1 | NP_001229417.1 | 21, 22 | 4912 | |||
NM_015037.3 | NP_055852.2 | 21, 22 | 4927 | |||
XM_005269648.2 | XP_005269705.1 | 21, 22 | 4974 | |||
XM_005269653.3 | XP_005269710.1 | 21, 22 | 4672 | |||
XM_005269655.3 | XP_005269712.1 | 22, 23 | 4570 | |||
XM_006717725.3 | XP_006717788.1 | 21, 22 | 4959 | |||
XM_017015978.1 | XP_016871467.1 | 21, 22 | 4959 | |||
GenBank mRNA | AB020720.2 | - | 22, 23 | 4813 | ||
AK023742.1 | - | 12, 13 | 2481 | |||
AL133662.1 | - | 13, 14 | 3354 | |||
BC040726.1 | - | 1, 2 | 194 | |||
BC111006.1 | - | 12, 13 | 2675 | |||
BC117205.1 | - | 12, 13 | 2474 | |||
BC151206.1 | - | 21, 22 | 4874 | 40147119 |
