siRNA Details
Gene Symbol: NEPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319111.1 | NP_001306040.1 | 3 | 366 | ||
NM_001319112.1 | NP_001306041.1 | 3 | 324 | |||
NM_001319114.1 | NP_001306043.1 | 3 | 370 | |||
NM_001319115.1 | NP_001306044.1 | 3 | 366 | |||
NM_015412.3 | NP_056227.2 | 4 | 496 | |||
XM_005247276.2 | XP_005247333.1 | 4 | 463 | |||
XM_011512635.2 | XP_011510937.1 | 4 | 459 | |||
XM_011512636.2 | XP_011510938.1 | 4 | 463 | |||
XM_011512638.2 | XP_011510940.1 | 3 | 358 | |||
XM_017006102.1 | XP_016861591.1 | 4 | 459 | |||
XM_017006103.1 | XP_016861592.1 | 3 | 354 | |||
GenBank mRNA | AK024325.1 | - | 4 | 454 | ||
AK295165.1 | - | 3 | 255 | |||
AK300811.1 | - | 3 | 299 | |||
AK301161.1 | - | 3 | 284 | |||
AK304083.1 | - | 3 | 299 | |||
AL117573.1 | - | 4 | 438 | |||
AL537045.3 | - | 3 | 291 | |||
BC014673.2 | - | 4 | 494 | |||
BC067743.1 | - | 4 | 456 | 4812681 |
