siRNA Details
Gene Symbol: ZNF385A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130967.2 | NP_001124439.1 | 8 | 1009 | ||
NM_001130968.2 | NP_001124440.1 | 7 | 766 | |||
NM_001290001.1 | NP_001276930.1 | 6 | 861 | |||
NM_001290002.1 | NP_001276931.1 | 9 | 1199 | |||
NM_001290004.1 | NP_001276933.1 | 9 | 1154 | |||
NM_015481.2 | NP_056296.1 | 7 | 1104 | |||
XM_005268783.4 | XP_005268840.1 | 8 | 1031 | |||
XM_006719342.2 | XP_006719405.1 | 9 | 1202 | |||
XM_011538168.2 | XP_011536470.1 | 9 | 1121 | |||
XM_011538169.2 | XP_011536471.1 | 8 | 1025 | |||
XM_011538170.1 | XP_011536472.1 | 7 | 874 | |||
XM_011538171.2 | XP_011536473.1 | 8 | 1169 | |||
XM_017019174.1 | XP_016874663.1 | 8 | 1337 | |||
GenBank mRNA | AB593085.1 | - | 9 | 1064 | ||
AF304052.1 | - | 7 | 1071 | |||
AK024404.1 | - | 7 | 742 | |||
AK092724.1 | - | 8 | 878 | |||
AK296564.1 | - | 6 | 804 | |||
AK315613.1 | - | 8 | 1002 | |||
AL117462.1 | - | 6 | 714 | |||
AY461717.1 | - | 9 | 1179 | |||
BC029752.1 | - | 8 | 990 | 4476647 | ||
CR457327.1 | - | 6 | 705 |
