siRNA Details
Gene Symbol: KIAA1429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015496.4 | NP_056311.2 | 8 | 2075 | ||
NM_183009.2 | NP_892121.1 | 8 | 2075 | |||
XM_017013306.1 | XP_016868795.1 | 7 | 1855 | |||
XM_017013307.1 | XP_016868796.1 | 7 | 1898 | |||
XM_017013308.1 | XP_016868797.1 | 6 | 1888 | |||
GenBank mRNA | AB037850.1 | - | 8 | 1999 | ||
AL832487.1 | - | 8 | 2016 | |||
BC069239.1 | - | 1 | 140 | |||
BC112288.1 | - | 8 | 2087 | 8327756 | ||
BC113380.1 | - | 8 | 2075 | 8322432 |
