siRNA Details
Gene Symbol: ZNF451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031623.2 | NP_001026794.1 | 6 | 782 | ||
NM_015555.2 | NP_056370.2 | 6 | 782 | |||
XM_005248994.1 | XP_005249051.1 | 6 | 802 | |||
XM_011514460.1 | XP_011512762.1 | 5 | 541 | |||
XM_011514461.1 | XP_011512763.1 | 4 | 1157 | |||
XM_011514462.2 | XP_011512764.1 | 6 | 802 | |||
GenBank mRNA | AB011148.1 | - | 6 | 601 | ||
AK225806.1 | - | 6 | 609 | |||
AY055204.1 | - | 6 | 783 | |||
BC021712.2 | - | 6 | 699 | 4821000 | ||
BC098382.1 | - | 6 | 625 | 5295845 | ||
BX648081.1 | - | 6 | 628 | |||
CN314846.1 | - | 2 | 185 |
