siRNA Details
Gene Symbol: SZRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114600.2 | NP_001108072.1 | 3, 4 | 595 | ||
NM_001271869.1 | NP_001258798.1 | 3, 4 | 592 | |||
NR_073500.1 | 1, 2 | 290 | ||||
NR_073501.1 | 4, 5 | 666 | ||||
NR_073502.1 | 2, 3 | 545 | ||||
NR_073503.1 | 2, 3 | 542 | ||||
GenBank mRNA | AB097042.1 | - | 2, 3 | 333 | ||
AB097043.1 | - | 2, 3 | 333 | |||
AB097044.1 | - | 2, 3 | 324 | |||
AB097045.1 | - | 2, 3 | 324 | |||
AB097046.1 | - | 3, 4 | 383 | |||
AJ295987.1 | - | 2, 3 | 329 | |||
AK297926.1 | - | 3, 4 | 383 | |||
AK298576.1 | - | 2, 3 | 329 | |||
AK308787.1 | - | 1, 2 | 77 | |||
AK316205.1 | - | 2, 3 | 331 | |||
AL050028.1 | - | 2, 3 | 329 | |||
BC023988.1 | - | 2, 3 | 350 | 3825796 | ||
BC065538.1 | - | 2, 3 | 330 | 6053254 | ||
DA010145.1 | - | 3, 4 | 387 | |||
DA438597.1 | - | 4, 5 | 453 |
