siRNA Details
Gene Symbol: FBXW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012164.3 | NP_036296.2 | 4, 5 | 928 | ||
XM_005251910.3 | XP_005251967.1 | 4, 5 | 931 | |||
XM_005251913.2 | XP_005251970.1 | 4, 5 | 830 | |||
XM_006717049.3 | XP_006717112.1 | 4, 5 | 1113 | |||
XM_011518518.1 | XP_011516820.1 | 4, 5 | 1168 | |||
XM_017014616.1 | XP_016870105.1 | 4, 5 | 1027 | |||
XM_017014617.1 | XP_016870106.1 | 4, 5 | 1051 | |||
GenBank mRNA | AF129531.1 | - | 2, 3 | 690 | ||
AF145024.1 | - | 4, 5 | 835 | |||
AF176698.1 | - | 3, 4 | 813 | |||
AK022484.1 | - | 4, 5 | 878 | |||
AK300066.1 | - | 4, 5 | 759 | |||
AK314385.1 | - | 4, 5 | 1113 | |||
BC018738.1 | - | 4, 5 | 844 | 4554961 | ||
BC110334.1 | - | 3, 4 | 828 | 5741924 |
