siRNA Details
Gene Symbol: OR1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005234.1 | NP_001005234.1 | 1 | 21 | ||
GenBank mRNA | BC136857.1 | - | 1 | 115 | 9020848 | |
BC136859.1 | - | 1 | 115 | 9020850 |
