siRNA Details
Gene Symbol: SLC39A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271957.1 | NP_001258886.1 | 4 | 1074 | ||
NM_001271958.1 | NP_001258887.1 | 4 | 911 | |||
NM_001271959.1 | NP_001258888.1 | 4 | 1080 | |||
NM_001271960.1 | NP_001258889.1 | 5 | 972 | |||
NM_001271961.1 | NP_001258890.1 | 3 | 780 | |||
NM_014437.4 | NP_055252.2 | 5 | 1218 | |||
GenBank mRNA | AF151829.1 | - | 4 | 789 | ||
AJ271671.1 | - | 4 | 819 | |||
AK074943.1 | - | 4 | 1028 | |||
AK075257.1 | - | 4 | 842 | |||
AK293389.1 | - | 3 | 688 | |||
AK297983.1 | - | 5 | 786 | |||
AK300244.1 | - | 5 | 778 | |||
BC002563.2 | - | 4 | 793 | 3138575 | ||
BC003152.2 | - | 3 | 765 | 2958651 | ||
BC007886.2 | - | 4 | 790 | 4122086 | ||
BC014303.2 | - | 5 | 1198 | 3942586 | ||
CR457217.1 | - | 3 | 732 |
