siRNA Details
Gene Symbol: UBIAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013319.2 | NP_037451.1 | 2 | 1104 | ||
GenBank mRNA | AF117064.1 | - | 2 | 1102 | ||
AK074890.1 | - | 2 | 1096 | |||
AK222808.1 | - | 2 | 1104 | |||
BC004468.2 | - | 2 | 796 | 3936638 | ||
BT006832.1 | - | 2 | 770 |
