siRNA Details
Gene Symbol: CCDC53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301107.1 | NP_001288036.1 | 4 | 416 | ||
NM_016053.3 | NP_057137.1 | 4 | 416 | |||
XM_011538423.1 | XP_011536725.1 | 4 | 334 | |||
XM_017019383.1 | XP_016874872.1 | 3 | 231 | |||
GenBank mRNA | AF151874.1 | - | 4 | 361 | ||
AF155655.1 | - | 4 | 361 | |||
AK223339.1 | - | 4 | 352 | |||
AK314970.1 | - | 4 | 334 | |||
BC010889.1 | - | 4 | 287 | 4272385 | ||
BX356987.2 | - | 4 | 371 | |||
CR457171.1 | - | 4 | 254 | |||
DB452335.1 | - | 4 | 416 |
