siRNA Details
Gene Symbol: BFAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016561.2 | NP_057645.1 | 7 | 1308 | ||
XM_005255350.2 | XP_005255407.1 | 6 | 1083 | |||
XM_011522520.1 | XP_011520822.1 | 5 | 913 | |||
GenBank mRNA | AF173003.1 | - | 7 | 1180 | ||
AK291114.1 | - | 7 | 1116 | |||
AK300778.1 | - | 5 | 881 | |||
AK313890.1 | - | 6 | 1116 | |||
BC003054.1 | - | 7 | 1139 | 3537783 |
