siRNA Details
Gene Symbol: LIPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204830.1 | NP_001191759.1 | 3 | 1043 | ||
NM_015929.3 | NP_057013.1 | 3 | 845 | |||
NM_145197.2 | NP_660198.1 | 4 | 973 | |||
NM_145198.2 | NP_660199.1 | 4 | 961 | |||
NM_145199.2 | NP_660200.1 | 2 | 719 | |||
NR_037935.1 | 4 | 1106 | ||||
NR_037936.1 | 3 | 782 | ||||
GenBank mRNA | AB017566.1 | - | 3 | 845 | ||
BC007001.1 | - | 3 | 765 | 3961227 | ||
BC009772.1 | - | 3 | 786 | 4091817 | ||
BC070145.1 | - | 3 | 823 | 4671058 | ||
BQ422048.1 | - | 3 | 754 |
