siRNA Details
Gene Symbol: TMEM104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017728.3 | NP_060198.3 | 9, 10 | 882 | ||
XM_006721963.1 | XP_006722026.1 | 9, 10 | 853 | |||
XM_017024797.1 | XP_016880286.1 | 9, 10 | 821 | |||
GenBank mRNA | AK074029.1 | - | 9, 10 | 842 | ||
AK291159.1 | - | 9, 10 | 877 | |||
BC037278.1 | - | 9, 10 | 834 | 4825529 |
