siRNA Details
Gene Symbol: MCUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017918.4 | NP_060388.2 | 8 | 1104 | ||
XM_006714246.3 | XP_006714309.1 | 8 | 1215 | |||
GenBank mRNA | AK000654.1 | - | 6 | 791 | ||
AK095936.1 | - | 8 | 1011 | |||
AK291098.1 | - | 8 | 1098 | |||
AL832311.1 | - | 3 | 975 | |||
BC002633.2 | - | 8 | 1042 | 3613298 | ||
BM830891.1 | - | 4 | 478 |
