siRNA Details
Gene Symbol: LINC00483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073199.1 | 2 | 96 | |||
GenBank mRNA | AK000670.1 | - | 2 | 97 | ||
AK000701.1 | - | 1 | 552 | |||
AK222717.1 | - | 1 | 550 | |||
AK310037.1 | - | 2 | 138 | |||
BC128165.1 | - | 1 | 451 | |||
BC128166.1 | - | 1 | 451 | |||
BP382918.1 | - | 2 | 96 |
