siRNA Details
Gene Symbol: BANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083907.2 | NP_001077376.2 | 6 | 975 | ||
NM_001127507.2 | NP_001120979.2 | 5 | 807 | |||
NM_017935.4 | NP_060405.4 | 6 | 1206 | |||
XM_017008337.1 | XP_016863826.1 | 6 | 953 | |||
GenBank mRNA | AB063170.1 | - | 5 | 842 | ||
AK000713.1 | - | 6 | 922 | |||
AK091523.1 | - | 10 | 1307 | |||
AK292133.1 | - | 6 | 1050 | |||
AL834291.1 | - | 6 | 1025 | |||
BC032241.1 | - | 6 | 1074 | 5214147 | ||
EU051376.1 | - | 5 | 545 |
