siRNA Details
Gene Symbol: WDR70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018034.2 | NP_060504.1 | 9 | 956 | ||
XM_005248318.3 | XP_005248375.1 | 9 | 974 | |||
XM_011514061.2 | XP_011512363.1 | 3 | 269 | |||
XM_017009604.1 | XP_016865093.1 | 8 | 790 | |||
XM_017009605.1 | XP_016865094.1 | 8 | 757 | |||
GenBank mRNA | AK001095.1 | - | 9 | 898 | ||
AK291179.1 | - | 9 | 978 | |||
AK310901.1 | - | 9 | 981 | |||
AL512685.1 | - | 9 | 957 | |||
BC009648.1 | - | 9 | 956 | 3454326 | ||
BC025315.1 | - | 9 | 934 | 5458850 | ||
BX537655.1 | - | 8 | 913 |
