siRNA Details
Gene Symbol: C14orf105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283056.1 | NP_001269985.1 | 4 | 617 | ||
NM_001283057.1 | NP_001269986.1 | 4 | 617 | |||
NM_001283058.1 | NP_001269987.1 | 4 | 664 | |||
NM_001283059.1 | NP_001269988.1 | 4 | 644 | |||
NM_001283060.1 | NP_001269989.1 | 4 | 597 | |||
NM_018168.3 | NP_060638.2 | 4 | 620 | |||
XM_005267806.1 | XP_005267863.1 | 4 | 613 | |||
XM_005267810.3 | XP_005267867.1 | 4 | 714 | |||
XM_005267811.3 | XP_005267868.1 | 3 | 292 | |||
XM_005267813.1 | XP_005267870.1 | 4 | 689 | |||
XM_005267814.1 | XP_005267871.1 | 4 | 686 | |||
XM_006720188.2 | XP_006720251.1 | 4 | 613 | |||
XM_006720189.2 | XP_006720252.1 | 4 | 333 | |||
GenBank mRNA | AI733133.1 | - | 3 | 419 | ||
AK001512.1 | - | 4 | 609 | |||
AK223127.1 | - | 4 | 608 | |||
AK307238.1 | - | 4 | 680 | |||
BC028060.1 | - | 4 | 592 | |||
BC117405.1 | - | 4 | 511 | 40125956 | ||
BC143565.1 | - | 4 | 558 | |||
BC143566.1 | - | 4 | 511 | 9052077 | ||
BC143567.1 | - | 4 | 511 | 9052078 | ||
BC143568.1 | - | 4 | 538 | |||
BI760135.1 | - | 4 | 591 |
