siRNA Details
Gene Symbol: KIAA1551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018169.3 | NP_060639.3 | 6 | 5639 | ||
XM_005253405.2 | XP_005253462.1 | 5 | 6090 | |||
XM_011520722.1 | XP_011519024.1 | 6 | 5799 | |||
XM_017019550.1 | XP_016875039.1 | 5 | 6499 | |||
GenBank mRNA | AK001514.1 | - | 3 | 1123 | ||
BC098115.1 | - | 3 | 1103 | |||
BC114509.1 | - | 3 | 1103 | |||
BC114956.1 | - | 3 | 1103 | |||
BC140819.1 | - | 6 | 5590 | 9021687 | ||
BM711484.1 | - | 3 | 356 | |||
CA419041.1 | - | 1 | 604 |
