siRNA Details
Gene Symbol: AP5S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204446.1 | NP_001191375.1 | 3 | 734 | ||
NM_001204447.1 | NP_001191376.1 | 3 | 582 | |||
NM_018347.2 | NP_060817.1 | 3 | 638 | |||
GenBank mRNA | AK002030.1 | - | 3 | 606 | ||
AK093332.1 | - | 3 | 550 | |||
BC043344.1 | - | 3 | 575 | 5505030 |
