siRNA Details
Gene Symbol: ZNF692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136036.2 | NP_001129508.1 | 11, 12 | 1685 | ||
NM_001193328.1 | NP_001180257.1 | 10, 11 | 1237 | |||
NM_017865.3 | NP_060335.2 | 11, 12 | 1469 | |||
XM_011544220.1 | XP_011542522.1 | 11, 12 | 1726 | |||
XM_011544221.1 | XP_011542523.1 | 11, 12 | 1723 | |||
XM_011544222.1 | XP_011542524.1 | 11, 12 | 1341 | |||
XM_011544223.2 | XP_011542525.1 | 10, 11 | 1591 | |||
XM_011544227.1 | XP_011542529.1 | 11, 12 | 1637 | |||
XM_017001698.1 | XP_016857187.1 | 11, 12 | 1338 | |||
XM_017001699.1 | XP_016857188.1 | 10, 11 | 1648 | |||
XM_017001700.1 | XP_016857189.1 | 10, 11 | 1263 | |||
XM_017001701.1 | XP_016857190.1 | 11, 12 | 1453 | |||
XM_017001702.1 | XP_016857191.1 | 10, 11 | 1378 | |||
XM_017001703.1 | XP_016857192.1 | 11, 12 | 1656 | |||
XM_017001704.1 | XP_016857193.1 | 11, 12 | 1613 | |||
XM_017001705.1 | XP_016857194.1 | 11, 12 | 1516 | |||
GenBank mRNA | AB209706.1 | - | 7, 8 | 1523 | ||
AF256175.1 | - | 4, 5 | 786 | |||
AK000538.1 | - | 10, 11 | 1206 | |||
AK001070.1 | - | 9, 10 | 885 | |||
AK225535.1 | - | 10, 11 | 1206 | |||
AK302187.1 | - | 11, 12 | 1619 | |||
AL133100.1 | - | 9, 10 | 2198 | |||
BC002948.1 | - | 11, 12 | 1425 | 3533691 |
