siRNA Details
Gene Symbol: RADIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018059.4 | NP_060529.4 | 4 | 1226 | ||
GenBank mRNA | AB058752.1 | - | 4 | 1170 | ||
AK001186.1 | - | 1 | 209 | |||
AK294745.1 | - | 4 | 1186 | |||
AK296325.1 | - | 3 | 747 | |||
AK302549.1 | - | 1218 | ||||
BC117317.1 | - | 3 | 1112 | 40125868 | ||
BC126311.1 | - | 3 | 1112 | 8992027 | ||
BC143526.1 | - | 3 | 1112 | 9052035 | ||
EF560723.1 | - | 3 | 1135 |
