siRNA Details
Gene Symbol: ANGPTL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018687.6 | NP_061157.3 | 1 | 235 | ||
GenBank mRNA | AF271350.2 | - | 3 | 586 | ||
AY358356.1 | - | 1 | 219 | |||
BP316755.1 | - | 1 | 236 | |||
KF809856.1 | - | 1 | 153 | |||
KF809858.1 | - | 1 | 153 |
