siRNA Details
Gene Symbol: LANCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018697.3 | NP_061167.1 | 7 | 1640 | ||
GenBank mRNA | AB035966.1 | - | 7 | 1117 | ||
AF353942.1 | - | 7 | 1117 | |||
AJ278245.1 | - | 7 | 1248 | |||
AK095857.1 | - | 7 | 1640 | |||
AK225958.1 | - | 7 | 1093 | |||
AK301747.1 | - | 7 | 1046 | |||
AK313326.1 | - | 7 | 1385 | |||
AL713665.2 | - | 7 | 1032 | |||
BC004887.2 | - | 4 | 468 | |||
BC070049.1 | - | 7 | 1336 | 30343188 |
