siRNA Details
Gene Symbol: CD177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020406.3 | NP_065139.2 | 3 | 388 | ||
XM_017027021.1 | XP_016882510.1 | 3 | 372 | |||
XM_017027022.1 | XP_016882511.1 | 3 | 372 | |||
GenBank mRNA | AB237911.1 | - | 3 | 340 | ||
AF146747.1 | - | 3 | 367 | |||
AJ290452.1 | - | 3 | 359 | |||
AJ305326.1 | - | 3 | 332 | |||
AJ310433.1 | - | 3 | 359 | |||
AK225226.1 | - | 3 | 361 | |||
AK290965.1 | - | 3 | 363 | |||
AM884905.1 | - | 3 | 356 | |||
AY358932.1 | - | 3 | 348 | |||
BC029167.1 | - | 3 | 350 | 5182826 | ||
BT020111.1 | - | 3 | 332 | |||
CD514938.1 | - | 3 | 336 | |||
DA728784.1 | - | 3 | 388 |
