siRNA Details
Gene Symbol: KIAA1324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267048.1 | NP_001253977.1 | 13 | 2030 | ||
NM_001284352.1 | NP_001271281.1 | 14 | 2170 | |||
NM_001284353.1 | NP_001271282.1 | 8 | 2151 | |||
NM_020775.4 | NP_065826.2 | 15 | 2291 | |||
XM_011541825.1 | XP_011540127.1 | 15 | 2151 | |||
XM_011541826.2 | XP_011540128.1 | 15 | 2151 | |||
XM_011541827.1 | XP_011540129.1 | 14 | 2001 | |||
XM_017001883.1 | XP_016857372.1 | 14 | 2149 | |||
GenBank mRNA | AK026832.1 | - | 7 | 925 | ||
AK301598.1 | - | 12 | 1506 |
