siRNA Details
Gene Symbol: WDFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020830.4 | NP_065881.1 | 10 | 1084 | ||
GenBank mRNA | AB037856.1 | - | 10 | 997 | ||
AF411977.1 | - | 10 | 981 | |||
AJ310568.1 | - | 10 | 1071 | |||
AK022888.1 | - | 10 | 1010 | |||
AK023415.1 | - | 9 | 878 | |||
BC040525.1 | - | 10 | 996 | 5285567 | ||
BC065934.1 | - | 10 | 1028 | 6164567 |
