siRNA Details
Gene Symbol: VAT1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020927.2 | NP_065978.1 | 4 | 767 | ||
GenBank mRNA | AB046796.1 | - | 4 | 737 | ||
AK290153.1 | - | 4 | 745 | |||
AL834240.1 | - | 3 | 382 | |||
BC033537.1 | - | 4 | 745 | 4821265 |
