siRNA Details
Gene Symbol: RAB40C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172663.1 | NP_001166134.1 | 7 | 849 | ||
NM_001172664.1 | NP_001166135.1 | 7 | 741 | |||
NM_001172665.1 | NP_001166136.1 | 7 | 701 | |||
NM_001172666.1 | NP_001166137.1 | 7 | 644 | |||
NM_021168.4 | NP_066991.3 | 6 | 746 | |||
GenBank mRNA | AY823398.1 | - | 6 | 614 | ||
BC001581.1 | - | 3 | 340 | |||
BC028696.1 | - | 6 | 748 | 4826800 | ||
BQ685517.1 | - | 7 | 680 |
