siRNA Details
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021185.4 | NP_067008.3 | 15 | 1815 | ||
XM_005259114.4 | XP_005259171.1 | 15 | 1911 | |||
XM_005259115.4 | XP_005259172.1 | 15 | 1834 | |||
XM_005259116.2 | XP_005259173.1 | 13 | 1635 | |||
XM_006723308.3 | XP_006723371.1 | 15 | 2431 | |||
XM_006723310.2 | XP_006723373.1 | 15 | 1698 | |||
XM_006723311.2 | XP_006723374.1 | 15 | 1836 | |||
XM_011527174.2 | XP_011525476.1 | 15 | 1825 | |||
XM_011527175.2 | XP_011525477.1 | 15 | 1731 | |||
XM_011527176.2 | XP_011525478.1 | 15 | 1734 | |||
XM_011527177.2 | XP_011525479.1 | 15 | 1695 | |||
XM_011527178.2 | XP_011525480.1 | 16 | 2177 | |||
XM_011527179.1 | XP_011525481.1 | 15 | 1815 | |||
XM_011527182.2 | XP_011525484.1 | 14 | 1559 | |||
XM_017027053.1 | XP_016882542.1 | 14 | 1695 | |||
XM_017027054.1 | XP_016882543.1 | 13 | 1428 | |||
XM_017027055.1 | XP_016882544.1 | 2 | 904 | |||
GenBank mRNA | AK128220.1 | - | 15 | 1598 | ||
AK310460.1 | - | 11 | 1145 | |||
AL133023.1 | - | 15 | 2431 | |||
BC094785.1 | - | 15 | 1735 | |||
BC109035.1 | - | 11 | 1214 | 40028284 |
